D(+)-Xylose, 99+%
D(+)-Xylose, 99+%
D(+)-Xylose, 99+%
Thermo Scientific Chemicals

D(+)-Xylose, 99+%

CAS: 58-86-6 | C5H10O5 | 150.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 141005000
Price (USD)
158.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
158.00
Each
Chemical Identifiers
CAS58-86-6
IUPAC Nameoxane-2,3,4,5-tetrol
Molecular FormulaC5H10O5
InChI KeySRBFZHDQGSBBOR-UHFFFAOYNA-N
SMILESOC1COC(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Water=<0.2 % (K.F.)
Heavy metals (as Pb)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only