4-Aminobenzoic Acid, 99%
4-Aminobenzoic Acid, 99%
4-Aminobenzoic Acid, 99%
Thermo Scientific Chemicals

4-Aminobenzoic Acid, 99%

CAS: 150-13-0 | C7H7NO2 | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 146212500
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
56.40
Save 8.75
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
56.40
Save 8.75
Each
Chemical Identifiers
CAS150-13-0
IUPAC Name4-aminobenzoic acid
Molecular FormulaC7H7NO2
InChI KeyALYNCZNDIQEVRV-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Appearance (Color)White to light yellow to light grey to light brown
Melting point186°C to 189°C
Titration with NaOH98.5 to 101.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only