4-Nitroanisole, 99+%
4-Nitroanisole, 99+%
4-Nitroanisole, 99+%
Thermo Scientific Chemicals

4-Nitroanisole, 99+%

CAS: 100-17-4 | C7H7NO3 | 153.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
Packaging:
Glass bottle
Catalog number 147020050
Price (USD)
24.65
Online exclusive
27.60
Save 2.95
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
24.65
Online exclusive
27.60
Save 2.95
Each
Chemical Identifiers
CAS100-17-4
IUPAC Name1-methoxy-4-nitrobenzene
Molecular FormulaC7H7NO3
InChI KeyBNUHAJGCKIQFGE-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Melting point51°C to 53°C
Appearance (Color)Green to brown
Infrared spectrumConforms
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only