Triphenyl phosphate, 99+%
Triphenyl phosphate, 99+%
Triphenyl phosphate, 99+%
Triphenyl phosphate, 99+%
Thermo Scientific Chemicals

Triphenyl phosphate, 99+%

CAS: 115-86-6 | C18H15O4P | 326.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 147672500
Price (USD)
35.65
Special offer
Online exclusive
Ends: 28-Dec-2026
41.50
Save 5.85
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
35.65
Special offer
Online exclusive
Ends: 28-Dec-2026
41.50
Save 5.85
Each
Chemical Identifiers
CAS115-86-6
IUPAC Nametriphenyl phosphate
Molecular FormulaC18H15O4P
InChI KeyXZZNDPSIHUTMOC-UHFFFAOYSA-N
SMILESO=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Phenol=<0.005 % (Free phenols)
Phenol=<0.005 % (Free phenols)

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only