8-Hydroxyquinoline sulfate monohydrate, 98%
8-Hydroxyquinoline sulfate monohydrate, 98%
8-Hydroxyquinoline sulfate monohydrate, 98%
Thermo Scientific Chemicals

8-Hydroxyquinoline sulfate monohydrate, 98%

CAS: 207386-91-2 | 0·5 H2O | 203.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 148455000
Price (USD)
193.65
Special offer
Online exclusive
Ends: 30-Jun-2026
228.00
Save 34.35 (15%)
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
193.65
Special offer
Online exclusive
Ends: 30-Jun-2026
228.00
Save 34.35 (15%)
Each
Chemical Identifiers
CAS207386-91-2
IUPAC Namebis(quinolin-8-ol) sulfuric acid hydrate
Molecular FormulaC18H18N2O7S
InChI KeyBNCXJZDIJIVJJO-UHFFFAOYSA-N
SMILESO.OS(O)(=O)=O.OC1=C2N=CC=CC2=CC=C1.OC1=C2N=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Powder or crystals
Infrared spectrumConforms
HPLC>=97.5 %
Water=<5 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only