Cinnamyl alcohol, 98% trans
Cinnamyl alcohol, 98% trans
Cinnamyl alcohol, 98% trans
Thermo Scientific Chemicals

Cinnamyl alcohol, 98% trans

CAS: 104-54-1 | C9H10O | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
250 g
500 g
Packaging:
Glass bottle
Catalog number 148520050
Price (USD)
26.65
Special offer
Online exclusive
Ends: 28-Dec-2026
31.20
Save 4.55
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
26.65
Special offer
Online exclusive
Ends: 28-Dec-2026
31.20
Save 4.55
Each
Chemical Identifiers
CAS104-54-1
IUPAC Name(2Z)-3-phenylprop-2-en-1-ol
Molecular FormulaC9H10O
InChI KeyOOCCDEMITAIZTP-DAXSKMNVSA-N
SMILESOCC=C/C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point31°C to 35°C
Appearance (Color)White to light yellow
GC>=97.5 %
Appearance (Form)Fused crystalline solid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only