Acetanilide, 99+%
Acetanilide, 99+%
Acetanilide, 99+%
Thermo Scientific Chemicals

Acetanilide, 99+%

CAS: 103-84-4 | C8H9NO | 135.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
500 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 150810050
Price (USD)
34.65
Special offer
Online exclusive
Ends: 28-Dec-2026
40.00
Save 5.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
34.65
Special offer
Online exclusive
Ends: 28-Dec-2026
40.00
Save 5.35
Each
Chemical Identifiers
CAS103-84-4
IUPAC NameN-phenylacetamide
Molecular FormulaC8H9NO
InChI KeyFZERHIULMFGESH-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=99.0 %
Melting point111°C to 115°C
Appearance (Form)Powder or crystals or flakes
Appearance (Color)White to beige to grey to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only