N-N-Butyldiethanolamine, 98%
N-N-Butyldiethanolamine, 98%
N-N-Butyldiethanolamine, 98%
Thermo Scientific Chemicals

N-N-Butyldiethanolamine, 98%

CAS: 102-79-4 | C8H19NO2 | 161.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 154362500
Price (USD)
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS102-79-4
IUPAC Name2-[butyl(2-hydroxyethyl)amino]ethan-1-ol
Molecular FormulaC8H19NO2
InChI KeyGVNHOISKXMSMPX-UHFFFAOYSA-N
SMILESCCCCN(CCO)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.5 %
Appearance (Form)Clear liquid
Infrared spectrumConforms
Refractive index1.4610 to 1.4640 (20°C, 589 nm)
Color scale=<200 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only