N-Methyl-1,3-propanediamine, 99%
N-Methyl-1,3-propanediamine, 99%
N-Methyl-1,3-propanediamine, 99%
Thermo Scientific Chemicals

N-Methyl-1,3-propanediamine, 99%

CAS: 6291-84-5 | C4H12N2 | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Catalog number 155570010
Price (USD)
176.65
Special offer
Online exclusive
Ends: 28-Dec-2026
208.00
Save 31.35
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
176.65
Special offer
Online exclusive
Ends: 28-Dec-2026
208.00
Save 31.35
Each
Chemical Identifiers
CAS6291-84-5
IUPAC Name(3-aminopropyl)(methyl)amine
Molecular FormulaC4H12N2
InChI KeyQHJABUZHRJTCAR-UHFFFAOYSA-N
SMILESCNCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<50 APHA
GC>=98.5 %
Refractive index1.4458 to 1.4478 (20°C, 589 nm)
Water=<0.5 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only