1-Adamantanol, 99%
1-Adamantanol, 99%
1-Adamantanol, 99%
Thermo Scientific Chemicals

1-Adamantanol, 99%

CAS: 768-95-6 | C10H16O | 152.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 157100250
Price (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS768-95-6
IUPAC Nameadamantan-1-ol
Molecular FormulaC10H16O
InChI KeyVLLNJDMHDJRNFK-UHFFFAOYSA-N
SMILESOC12CC3CC(CC(C3)C1)C2
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Color)White to off-white
Infrared spectrumConforms
Appearance (Form)Crystalline powder or needles
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only