Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NK1.1 Antibodies

          Invitrogen

          NK1.1 Monoclonal Antibody (PK136), Functional Grade, eBioscience™

          57 Published Figures
          79 References
          View all (50) NK1.1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite NK1.1 Monoclonal Antibody (PK136), Functional Grade, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (57)
          NK1.1 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          NK1.1 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 58

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NK1.1 Antibody (16-5941-38) in Flow

          Staining of C57BL/6 mouse splenocytes with Anti-Mouse CD49b (Integrin alpha 2) FITC (Product # 11-0491-82) and Anti-Mouse NK-1.1 PE. Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Immunohistochemistry (IHC)
          NK1.1 Antibody in Immunohistochemistry (IHC)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)
          NK1.1 Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          16-5941-38

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Flow Cytometry (Flow)

          0.5 µg/test
          View 73 publications 73 publications

          Functional Assay (Functional)

          Assay-Dependent
          -

          Miscellaneous PubMed (Misc)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Mouse / IgG2a, kappa

          Recommended Isotype Control

          Mouse IgG2a kappa Isotype Control (eBM2a), Functional Grade, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          PK136

          Conjugate

          Functional Grade Functional Grade Functional Grade
          • Unconjugated
          • Alexa Fluor 561
          • Alexa Fluor 660 
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 496
          • Brilliant UV 615
          • Brilliant UV 661
          • Brilliant UV 805
          • Brilliant Violet 421
          • Brilliant Violet 480
          • Brilliant Violet 605
          • Brilliant Violet 650
          • Brilliant Violet 711
          • Brilliant Violet 786
          • eFluor 450
          • FITC
          • PE 
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-Cyanine5.5
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Super Bright 702
          • Super Bright 780
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          5 mg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          no preservative

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2866122

          Product Specific Information

          Description: The PK136 monoclonal antibody reacts with mouse NK1.1, an antigen expressed by natural killer cells and a subset of T cells in the NK1.1 mouse strains including C57BL and NZB. Several commonly used laboratory mouse strains such as BALB/c, SJL, AKR, CBA, C3H and A do not express the NK1.1 antigen. For detection of NK cells in these strains the monoclonal antibody DX5 (Product # 14-5971) should be used. Simultaneous staining of C57BL/6 spleen cells with PK136 and DX5 reveals coexpression of both markers by a majority of cells as well as presence of small populations of DX5+PK136- and DX5-PK136+ cells.

          Applications Reported: PK136 has been reported for use in flow cytometric analysis, depletion of NK cells, and in vitro functional studies.

          Applications Tested: The PK136 antibody has been tested by flow cytometric analysis of C57Bl/6 mouse splenocytes and can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in other applications.

          Storage and handling: Use in a sterile environment.

          Filtration: 0.2 µm post-manufacturing filtered.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Endotoxin Level: Less than 0.001 ng/µg antibody, as determined by LAL assay.

          Aggregation: Less than 10%, as determined by HPLC.

          Target Information

          NK1.1 (Killer cell lectin-like receptor subfamily B, member 1, KLRB1, NKR-P1A, CD161, cluster of differentiation 161), refers to Natural Killer (NK) cells, lymphocytes that mediate cytotoxicity and secrete cytokines after immune stimulation. Several genes of the C-type lectin superfamily, including the rodent NKRP1 family of glycoproteins, are expressed by NK cells and may be involved in the regulation of NK cell function. The KLRB1 protein contains an extracellular domain with several motifs characteristic of C-type lectins, a transmembrane domain, and a cytoplasmic domain. The KLRB1 protein, NKR-P1A or CD161, is classified as a type II membrane protein because it has an external C terminus. NKR-P1A, the receptor encoded by the KLRB1 gene, recognizes Lectin Like Transcript-1 (LLT1) as a functional ligand.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: C-type lectin-like receptor; CD161 antigen-like family member B; CD161 antigen-like family member C; inhibitory receptor NKR-P1B; killer cell lectin-like receptor subfamily A member 1C; killer cell lectin-like receptor subfamily B member 1D; KLRB1; Ly-55; Ly55; lymphocyte antigen 55 complex, locus B; lymphocyte antigen 55 complex, locus C; lymphocyte antigen 55 complex, locus D; lymphocyte antigen 55b; lymphocyte antigen 55c; lymphocyte antigen 55d; lymphocyte antigen 59; MGC163757; MGC163759; natural killer cell receptor protein NKR-P1B; natural killer cell receptor protein NKR-P1C; natural killer cell receptor-P1; natural killer cell surface protein NKR-P1B; natural killer cell surface protein P1-40; natural killer cell-associated antigen 1; natural killer cells; nk cells; NKR-P1 34; NKR-P1 40; NKR-P1 gene-34 protein; NKR-P1 gene-40 protein; NKR-P1B; receptor-like type II integral membrane protein; transmembrane protein; unnamed protein product

          View more View less

          Gene Aliases: CD161; Klrb1b; Klrb1d; ly-55b; ly-55c; ly-55d; Ly-59; Ly55b; Ly55c; Ly55d; Ly59; Nk-1; Nk-1.2; NK-RP1; Nk1; NK1.1; Nk1.2; NKR-P1.9; NKR-P1B; NKR-P1D; NKRP1; Nkrp1-b; NKRP140; Nkrp1b; Nkrp1c; Nkrp1d

          View more View less

          UniProt ID: (Mouse) Q99JB4, (Mouse) P27814

          View more View less

          Entrez Gene ID: (Mouse) 80782, (Mouse) 17059

          View more View less

          Function(s)
          protein binding signaling receptor activity identical protein binding
          Process(es)
          natural killer cell activation regulation of natural killer cell mediated cytotoxicity positive regulation of natural killer cell mediated cytotoxicity negative regulation of natural killer cell mediated cytotoxicity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-fr8qn:80/100.66.77.38:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0