Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • IL-22 Antibodies

          Invitrogen

          IL-22 Monoclonal Antibody (IL22JOP), Functional Grade, eBioscience™

          31 Published Figures
          45 References
          View all (72) IL-22 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite IL-22 Monoclonal Antibody (IL22JOP), Functional Grade, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (31)
          IL-22 Antibody in Functional Assay (Functional)
          Group 53 Created with Sketch.
          IL-22 Antibody in Functional Assay (Functional)
          Group 53 Created with Sketch.

          FIGURE: 1 / 32

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IL-22 Antibody (16-7222-025) in Functional

          Neutralization of IL-22 Recombinant Protein activity as measured by IL-10 induction in COLO205 cells. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IL-22 Antibody in Functional Assay (Functional)
          IL-22 Antibody in Immunohistochemistry (IHC)
          IL-22 Antibody in Flow Cytometry (Flow)
          IL-22 Antibody in Flow Cytometry (Flow)
          IL-22 Antibody in Flow Cytometry (Flow)
          IL-22 Antibody in Flow Cytometry (Flow)
          IL-22 Antibody in Flow Cytometry (Flow)
          IL-22 Antibody in Flow Cytometry (Flow)
          IL-22 Antibody in Flow Cytometry (Flow)
          IL-22 Antibody in Flow Cytometry (Flow)
          IL-22 Antibody in Flow Cytometry (Flow)
          IL-22 Antibody in Flow Cytometry (Flow)
          IL-22 Antibody in Flow Cytometry (Flow)
          IL-22 Antibody in Flow Cytometry (Flow)
          IL-22 Antibody in Flow Cytometry (Flow)
          IL-22 Antibody in Flow Cytometry (Flow)
          IL-22 Antibody in Flow Cytometry (Flow)
          IL-22 Antibody in Flow Cytometry (Flow)
          IL-22 Antibody in Flow Cytometry (Flow)
          IL-22 Antibody in Flow Cytometry (Flow)
          IL-22 Antibody in Flow Cytometry (Flow)
          IL-22 Antibody in Flow Cytometry (Flow)
          IL-22 Antibody in Flow Cytometry, Neutralization (Flow, Neu)
          IL-22 Antibody in Flow Cytometry, Neutralization (Flow, Neu)
          IL-22 Antibody in ELISA (ELISA)
          IL-22 Antibody in ELISA (ELISA)
          IL-22 Antibody in ELISA (ELISA)
          IL-22 Antibody in ELISA (ELISA)
          IL-22 Antibody in Neutralization (Neu)
          IL-22 Antibody in Neutralization (Neu)
          IL-22 Antibody in Neutralization (Neu)
          IL-22 Antibody in Functional Assay (Functional)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          16-7222-025

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Flow Cytometry (Flow)

          -
          View 26 publications 26 publications

          ELISA (ELISA)

          -
          View 2 publications 2 publications

          Neutralization (Neu)

          Assay-Dependent
          View 4 publications 4 publications

          Functional Assay (Functional)

          Assay-Dependent
          View 6 publications 6 publications

          Inhibition Assays (Inhibition)

          -
          View 1 publication 1 publication

          In vitro Assay (In vitro)

          -
          View 1 publication 1 publication

          Mass Spectrometry (MS)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rhesus monkey

          Published species

          Human, Mouse, Non-human primate, Rhesus monkey

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), Functional Grade, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          IL22JOP

          Conjugate

          Functional Grade Functional Grade Functional Grade
          • APC 
          • PerCP-eFluor 710
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          25 mg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          no preservative

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2866263

          Product Specific Information

          Description: The monoclonal antibody IL22JOP reacts with and inhibits the bioactivity of human and mouse IL-22. IL-22 is a 20 kDa member of the IL-10 cytokine family that is secreted primarily by Th17 cells, NK cells, and other T cells. Compared to IL-6 or TGF beta, IL-23 can induce greater levels of IL-22 in in vitro-differentiated Th17 cells. This observation suggests that IL-22 may be secreted by more fully differentiated Th17 cells in vivo. Recently, it was demonstrated that IL-22 could protect hosts from bacterial infection of the lungs and gut. Moreover, it has been reported that anti-CD3/CD28-induced production of IL-22 by PBMCs was elevated significantly in asthma patients compared to control patients. Flow cytometric analysis also showed that the frequencies of IL-17+IL-22+ CD4 T cells were increased in PBMCs from patients with ankylosing spondylitis and rheumatoid arthritis.

          IL22JOP is published to recognize rhesus IL-22.

          Applications Reported: The monoclonal antibody IL22JOP reacts with and inhibits the bioactivity of human and mouse IL-22.

          Applications Tested: The ND50 of IL22JOP, as determined by the inhibition of IL-10 induction in COLO205 cells by IL-22, is 10-40 ng/mL, in the presence of 0.2 ng/mL recombinant human or mouse IL-22. Neutralization dose will vary depending on assay method, cytokine concentration, and cell type.

          Storage and handling: Use in a sterile environment.

          Filtration: 0.2 µm post-manufacturing filtered.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Endotoxin Level: Less than 0.001 ng/µg antibody, as determined by LAL assay.

          Aggregation: Less than 10%, as determined by HPLC.

          Target Information

          IL-22 also known as IL-10-related T-cell derived inducible factor, is an alpha helical cytokine and is considered a member of the IFN-IL-10 family, which includes IL-19, IL-20, IL-24, IL-26, IL-28, IL-29, and the type I and II interferons. IL-22 is produced mainly by activated T cells and NK cells. In humans, the IL-22 gene is located on the q arm of chromosome 12, and is structurally related to IL10. IL-22 acts by engaging the heterodimeric receptor complex consisting of primary receptor IL-22R1 and accessory receptor IL-10R2. IL-22R1 also binds IL-20 and IL-24; IL-10R2 also binds IL-10, IL-27, IL-28, and IL-29. Binding of IL-22 to its receptor complex induces signal transduction, particularly via the JAK-STAT pathway. In addition to the membrane-bound IL-22R1/IL-10R2 complex, a soluble single-chain IL-22 receptor termed IL-22BP has been found to antagonize IL-22 binding and signaling. IL-22 appears not to directly influence immune cells, and major targets of the cytokine appear to be nonimmune cells, such as cells of the skin, digestive and respiratory system, as well as hepatocytes, and keratinocytes. IL-22 has been described as an effector cytokine of the Th17 lineage. Along with IL-17A and IL-17F, IL-22 regulates genes associated with innate immunity of the skin. IL-17A, IL-17F and IL-22 are all co-expressed by Th17 cells, however, they are differentially regulated. The effects of IL-22 include induction of acute phase reactants and antimicrobial proteins, as well as increasing the mobility of keratinocytes. IL-22 is highly expressed during chronic inflammation, and found to activate intracellular kinases and transcription factors. IL-22 is critical for host defense against infections of extracellular pathogens, and promotes wound-healing responses. IL-22 is upregulated in activated T cells. IL-22 has been reported to mediate IL-23-induced acanthosis and dermal inflammation through activation of STAT3.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: cytokine; cytokine Zcyto18; il 22; IL-10-related T-cell-derived inducible factor; IL-10-related T-cell-derived-inducible factor; IL-TIF alpha; ILN; ILTIF alpha; interferon 2b1; Interleukin; interleukin 10-related T cell-derived inducible factor; interleukin-22a; Interleukin22

          View more View less

          Gene Aliases: EGK_03903; If2b1; IL-21; IL-22; IL-22a; IL-D110; IL-TIF; ILTIF; ILTIFa; TIFa; TIFIL-23; zcyto18

          View more View less

          UniProt ID: (Human) Q9GZX6, (Mouse) Q9JJY9

          View more View less

          Entrez Gene ID: (Human) 50616, (Rhesus monkey) 718047, (Mouse) 50929

          View more View less

          Function(s)
          cytokine activity protein binding interleukin-22 receptor binding
          Process(es)
          acute-phase response inflammatory response signal transduction negative regulation of inflammatory response response to glucocorticoid regulation of tyrosine phosphorylation of STAT protein positive regulation of transcription from RNA polymerase II promoter reactive oxygen species metabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-8lbpj:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0