Donor Bovine Serum, New Zealand Origin
Gibco™

Donor Bovine Serum, New Zealand Origin

Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

Have Questions?
카탈로그 번호수량
16030074500 mL
카탈로그 번호 16030074
제품 가격(KRW)
109,375
온라인 행사
Ends: 30-Sep-2026
125,000
할인액 15,625 (13%)
Each
수량:
500 mL
제품 가격(KRW)
109,375
온라인 행사
Ends: 30-Sep-2026
125,000
할인액 15,625 (13%)
Each

Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

Sera Category: Secure

Origin: New Zealand.

Endotoxin level: ≤10 EU/ml.

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
사양
엔도톡신 농도≤ 10 EU/mL
제품라인Gibco
배송 조건Frozen
Cattle/Bovine
Age12 to 36 months
원산지New Zealand
형태Liquid
형식Bottle
제품 유형Calf Bovine Serum
수량500 mL
Serum TreatmentSterile-filtered
멸균Sterile-filtered
Unit SizeEach
구성 및 보관
Storage conditions: ≤-10°C
Shipping conditions: Frozen

자주 묻는 질문(FAQ)

Will improper storage of my tissue culture reagents affect the growth rate of my culture?

Yes. Store animal sera at -5 to -20 degrees C. Store media at 2 to 8 degrees C; use within recommended shelf life period. Store complete media (supplemented) at 2 to 8 degrees C, and for complete medium the recommended shelf life is 2 to 4 weeks. Additionally, minimize exposure of sera and media to light.