1-Pentanol, 99%, pure
1-Pentanol, 99%, pure
1-Pentanol, 99%, pure
Thermo Scientific Chemicals

1-Pentanol, 99%, pure

CAS: 71-41-0 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 160600025
Price (USD)
103.65
Online exclusive
115.00
Save 11.35
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
103.65
Online exclusive
115.00
Save 11.35
Each
Chemical Identifiers
CAS71-41-0
IUPAC Namepentan-1-ol
Molecular FormulaC5H12O
InChI KeyAMQJEAYHLZJPGS-UHFFFAOYSA-N
SMILESCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<20 APHA
Infrared spectrumConforms
Water=<0.2 %
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only