D(-)-Arabinose, 99+%
D(-)-Arabinose, 99+%
D(-)-Arabinose, 99+%
Thermo Scientific Chemicals

D(-)-Arabinose, 99+%

CAS: 28697-53-2 | C5H10O5 | 150.13 g/mol
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number 161450250
Price (JPY)
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS28697-53-2
IUPAC Name(2S,3R)-2,3,4,5-tetrahydroxypentanal
Molecular FormulaC5H10O5
InChI KeyPYMYPHUHKUWMLA-DEOSMSJNSA-N
SMILESOCC(O)[C@@H](O)[C@H](O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
Specific optical rotation-101.0° to 106.0° (20°C, 589 nm) (c=4, H2O)
Specific optical rotationafter 24 h
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only