p-Tolyl isocyanate, 99%
p-Tolyl isocyanate, 99%
p-Tolyl isocyanate, 99%
Thermo Scientific Chemicals

p-Tolyl isocyanate, 99%

CAS: 622-58-2 | C8H7NO | 133.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 162520050
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
39.90
Save 6.25
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
39.90
Save 6.25
Each
Chemical Identifiers
CAS622-58-2
IUPAC Name1-isocyanato-4-methylbenzene
Molecular FormulaC8H7NO
InChI KeyMGYGFNQQGAQEON-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5301 to 1.5321 (20°C, 589 nm)
Appearance (Color)Clear light brown
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only