Brucine dihydrate, 99%
Brucine dihydrate, 99%
Brucine dihydrate, 99%
Thermo Scientific Chemicals

Brucine dihydrate, 99%

CAS: 145428-94-0 | C23H27N2O4 | 395.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
Packaging:
Glass bottle
Catalog number 163501000
Price (EUR)
109,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
109,00
Each
Chemical Identifiers
CAS145428-94-0
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=98.5 % (On dry substance)
Appearance (Form)Powder or crystals
Melting point174°C to 179°C
Loss on drying6.0 % to 9.5 % (105°C, 3 hrs)
Appearance (Color)White to light beige
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only