1,3,5-Benzenetricarboxylic acid chloride, 98%
1,3,5-Benzenetricarboxylic acid chloride, 98%
1,3,5-Benzenetricarboxylic acid chloride, 98%
Thermo Scientific Chemicals

1,3,5-Benzenetricarboxylic acid chloride, 98%

CAS: 4422-95-1 | C9H3Cl3O3 | 265.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 164375000
Price (USD)
629.65
Special offer
Online exclusive
Ends: 28-Dec-2026
741.00
Save 111.35
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
629.65
Special offer
Online exclusive
Ends: 28-Dec-2026
741.00
Save 111.35
Each
Chemical Identifiers
CAS4422-95-1
IUPAC Namebenzene-1,3,5-tricarbonyl trichloride
Molecular FormulaC9H3Cl3O3
InChI KeyUWCPYKQBIPYOLX-UHFFFAOYSA-N
SMILESClC(=O)C1=CC(=CC(=C1)C(Cl)=O)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration Argentometric97.5 to 102.5 % (ex Cl)
Melting point30°C to 35°C
Appearance (Color)White to yellow
Appearance (Form)Solid or liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only