2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%
2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%
2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%
Thermo Scientific Chemicals

2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%

CAS: 77-99-6 | C6H14O3 | 134.18 g/mol
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic drum
Catalog number 164650100
Price (JPY)
-
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS77-99-6
IUPAC Name2-ethyl-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC6H14O3
InChI KeyZJCCRDAZUWHFQH-UHFFFAOYSA-N
SMILESCCC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Flakes
Melting point>=57°C
Ash=<0.0015 %
Acidity=<0.002 % (as Formic acid)
Water=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only