4-Methyl-3-nitrobenzoic acid, 99%
4-Methyl-3-nitrobenzoic acid, 99%
4-Methyl-3-nitrobenzoic acid, 99%
Thermo Scientific Chemicals

4-Methyl-3-nitrobenzoic acid, 99%

CAS: 96-98-0 | C8H7NO4 | 181.15 g/mol
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 165991000
Price (JPY)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS96-98-0
IUPAC Name4-methyl-3-nitrobenzoic acid
Molecular FormulaC8H7NO4
InChI KeyBBEWSMNRCUXQRF-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1[N+]([O-])=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to light yellow
Melting point187°C to 190°C
Appearance (Form)Crystalline powder
Titration with NaOH>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only