3-Hydroxy-4-nitrobenzaldehyde, 97%
3-Hydroxy-4-nitrobenzaldehyde, 97%
3-Hydroxy-4-nitrobenzaldehyde, 97%
Thermo Scientific Chemicals

3-Hydroxy-4-nitrobenzaldehyde, 97%

CAS: 704-13-2 | C7H5NO4 | 167.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number 168560010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS704-13-2
IUPAC Name3-hydroxy-4-nitrobenzaldehyde
Molecular FormulaC7H5NO4
InChI KeyAUBBVPIQUDFRQI-UHFFFAOYSA-N
SMILESOC1=CC(C=O)=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point130°C to 135°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only