Iron(III) chloride, 98%, pure, anhydrous
Iron(III) chloride, 98%, pure, anhydrous
Iron(III) chloride, 98%, pure, anhydrous
Thermo Scientific Chemicals

Iron(III) chloride, 98%, pure, anhydrous

CAS: 7705-08-0 | Cl3Fe | 162.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 kg
2.5 kg
10 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number 169430025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7705-08-0
IUPAC Nameiron(3+) trichloride
Molecular FormulaCl3Fe
InChI KeyRBTARNINKXHZNM-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Fe+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Manganese (Mn)=<3000 ppm
Appearance (Color)Dark grey to black
Titration Iodometric97.5 to 102.5 %
Copper (Cu)=<1000 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only