3-Methoxybenzylamine, 98%
3-Methoxybenzylamine, 98%
3-Methoxybenzylamine, 98%
Thermo Scientific Chemicals

3-Methoxybenzylamine, 98%

CAS: 5071-96-5 | C8H11NO | 137.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
25 mL
Packaging:
Glass bottle
Catalog number 170550100
Price (TWD)
-
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5071-96-5
IUPAC Name1-(3-methoxyphenyl)methanamine
Molecular FormulaC8H11NO
InChI KeyGRRIMVWABNHKBX-UHFFFAOYSA-N
SMILESCOC1=CC=CC(CN)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Refractive index1.5470 to 1.5490 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only