PIPES, 98.5+%, for biochemistry
PIPES, 98.5+%, for biochemistry
PIPES, 98.5+%, for biochemistry
Thermo Scientific Chemicals

PIPES, 98.5+%, for biochemistry

CAS: 5625-37-6 | C8H18N2O6S2 | 302.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 172610250
Price (USD)
44.60
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
44.60
Each
Chemical Identifiers
CAS5625-37-6
IUPAC Namesodium 2-[4-(2-sulfoethyl)piperazin-1-yl]ethane-1-sulfonate
Molecular FormulaC8H17N2NaO6S2
InChI KeyOGGAIRCLBMGXCZ-UHFFFAOYSA-M
SMILES[Na+].OS(=O)(=O)CCN1CCN(CCS([O-])(=O)=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<0.5 %
UV(0.1 M in 0.1 M NaOH) 1 cm cell
UVat 280 nm A: =<0.04
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only