Thermo Scientific Chemicals

Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals

Catalog number: 172805000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals

Catalog number: 172805000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 172805000
Price (USD)
Price: 685.00
Online price: 582.65
Your price:
Quantity
-

Chemical Identifiers

CAS
375-22-4
IUPAC Name
heptafluorobutanoic acid
Molecular Formula
C4HF7O2
InChI Key
YPJUNDFVDDCYIH-UHFFFAOYSA-N
SMILES
OC(=O)C(F)(F)C(F)(F)C(F)(F)F
Titration with NaOH
>=98.5 %
Infrared spectrum
Conforms
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text