3-Nitrobenzonitrile, 98%
3-Nitrobenzonitrile, 98%
3-Nitrobenzonitrile, 98%
Thermo Scientific Chemicals

3-Nitrobenzonitrile, 98%

CAS: 619-24-9 | C7H4N2O2 | 148.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 173520050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS619-24-9
IUPAC Name3-nitrobenzonitrile
Molecular FormulaC7H4N2O2
InChI KeyRUSAWEHOGCWOPG-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC(=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Form)Crystalline powder or needles
Melting point115°C to 119°C
Appearance (Color)Yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only