4,4'-Biphenol, 97%
4,4'-Biphenol, 97%
4,4'-Biphenol, 97%
Thermo Scientific Chemicals

4,4'-Biphenol, 97%

CAS: 92-88-6 | C12H10O2 | 186.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
250 g
Packaging:
View all
Plastic bottle
Catalog number 173671000
Price (USD)
268.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
268.00
Each
Chemical Identifiers
CAS92-88-6
IUPAC Name[1,1'-biphenyl]-4,4'-diol
Molecular FormulaC12H10O2
InChI KeyVCCBEIPGXKNHFW-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light beige
Infrared spectrumConforms
Appearance (Form)Powder
GC>=96.0 %
Melting point278°C to 283°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only