Tetrabutylammonium chloride, 95%, Tech., contains also bromide
Tetrabutylammonium chloride, 95%, Tech., contains also bromide
Tetrabutylammonium chloride, 95%, Tech., contains also bromide
Thermo Scientific Chemicals

Tetrabutylammonium chloride, 95%, Tech., contains also bromide

CAS: 1112-67-0 | C16H36ClN | 277.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 174120050
Price (USD)
32.65
Online Exclusive
35.60
Save 2.95 (8%)
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
32.65
Online Exclusive
35.60
Save 2.95 (8%)
Each
Chemical Identifiers
CAS1112-67-0
IUPAC Nametetrabutylazanium chloride
Molecular FormulaC16H36ClN
InChI KeyNHGXDBSUJJNIRV-UHFFFAOYSA-M
SMILES[Cl-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Appearance (Form)chunks or fused solid
Titration Argentometric>=94 % (On dry substance)
Bromide (Br)=<5 %
Appearance (Form)Crystalline powder or crystals or
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only