Ethylene glycol diacetate, 99%
Ethylene glycol diacetate, 99%
Ethylene glycol diacetate, 99%
Thermo Scientific Chemicals

Ethylene glycol diacetate, 99%

CAS: 111-55-7 | C6H10O4 | 146.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 176220025
Price (USD)
114.65
Online Exclusive
127.00
Save 12.35 (10%)
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
114.65
Online Exclusive
127.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS111-55-7
SpecificationsSpecification SheetSpecification Sheet
Acidity (CH3COOH)=<0.1 %
Appearance (Form)Clear liquid
Color scale=<10 APHA
Infrared spectrumConforms
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only