4-Bromophenethyl alcohol, 99%
4-Bromophenethyl alcohol, 99%
4-Bromophenethyl alcohol, 99%
Thermo Scientific Chemicals

4-Bromophenethyl alcohol, 99%

CAS: 4654-39-1 | C8H9BrO | 201.06 g/mol
Have Questions?
Catalog NumberQuantity
17959010010 g
Catalog number 179590100
Price (USD)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS4654-39-1
IUPAC Name2-(4-bromophenyl)ethan-1-ol
Molecular FormulaC8H9BrO
InChI KeyPMOSJSPFNDUAFY-UHFFFAOYSA-N
SMILESOCCC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear colorless to light yellow
Refractive index1.5720 to 1.5740 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only