5-Methoxy-1-indanone, 98%
5-Methoxy-1-indanone, 98%
5-Methoxy-1-indanone, 98%
Thermo Scientific Chemicals

5-Methoxy-1-indanone, 98%

CAS: 5111-70-6 | C10H10O2 | 162.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
Catalog number 179670050
Price (EUR)
73,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
73,00
Each
Chemical Identifiers
CAS5111-70-6
IUPAC Name5-methoxy-2,3-dihydro-1H-inden-1-one
Molecular FormulaC10H10O2
InChI KeyQOPRWBRNMPANKN-UHFFFAOYSA-N
SMILESCOC1=CC=C2C(=O)CCC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green-brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point107°C to 111°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only