Thermo Scientific Chemicals

2-Methoxyethanol, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 180790250
25 L, Metal drum, Each
Thermo Scientific Chemicals

2-Methoxyethanol, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 180790250
25 L, Metal drum, Each
Quantity
Packaging

Chemical Identifiers

CAS
109-86-4
IUPAC Name
2-methoxyethan-1-ol
Molecular Formula
C3H8O2
InChI Key
XNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILES
COCCO
Appearance (Form)
Clear liquid
Color scale
=<15 APHA
Infrared spectrum
Conforms
Residue after evaporation
=<0.005 %
GC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text