2-Bromobenzyl bromide, 98%
2-Bromobenzyl bromide, 98%
2-Bromobenzyl bromide, 98%
Thermo Scientific Chemicals

2-Bromobenzyl bromide, 98%

CAS: 3433-80-5 | C7H6Br2 | 249.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 181791000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3433-80-5
IUPAC Name1-bromo-2-(bromomethyl)benzene
Molecular FormulaC7H6Br2
InChI KeyLZSYGJNFCREHMD-UHFFFAOYSA-N
SMILESBrCC1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive index1.6170 to 1.6190 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid after melting
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only