3-Nitrobenzamide, 98%
3-Nitrobenzamide, 98%
3-Nitrobenzamide, 98%
Thermo Scientific Chemicals

3-Nitrobenzamide, 98%

CAS: 645-09-0 | C7H6N2O3 | 166.14 g/mol
製品番号(カタログ番号)数量パッケージ
18344025025 gGlass bottle
製品番号(カタログ番号) 183440250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS645-09-0
IUPAC Name3-nitrobenzamide
Molecular FormulaC7H6N2O3
InChI KeyKWAYEPXDGHYGRW-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=CC(=C1)[N+]([O-])=O
さらに表示
Appearance (Color)Yellow
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Crystalline powder or crystals
Melting point140°C to 144°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only