3,5-Bis(trifluoromethyl)aniline, 98+%
3,5-Bis(trifluoromethyl)aniline, 98+%
3,5-Bis(trifluoromethyl)aniline, 98+%
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)aniline, 98+%

CAS: 328-74-5 | C8H5F6N | 229.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Catalog number 185690100
Price (USD)
-
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS328-74-5
IUPAC Name3,5-bis(trifluoromethyl)aniline
Molecular FormulaC8H5F6N
InChI KeyCDIDGWDGQGVCIB-UHFFFAOYSA-N
SMILESNC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=98.0 %
Refractive index1.4330 to 1.4350 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear light yellow to yellow-brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only