2-Chlorophenethylalcohol, 98%
2-Chlorophenethylalcohol, 98%
2-Chlorophenethylalcohol, 98%
Thermo Scientific Chemicals

2-Chlorophenethylalcohol, 98%

CAS: 19819-95-5 | C8H9ClO | 156.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 186190050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS19819-95-5
IUPAC Name2-(2-chlorophenyl)ethan-1-ol
Molecular FormulaC8H9ClO
InChI KeyIWNHTCBFRSCBQK-UHFFFAOYSA-N
SMILESOCCC1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.5495 to 1.5515 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only