Chromium(III) acetylacetonate, 97%
Chromium(III) acetylacetonate, 97%
Chromium(III) acetylacetonate, 97%
Thermo Scientific Chemicals

Chromium(III) acetylacetonate, 97%

CAS: 21679-31-2 | C15H21CrO6 | 349.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
View all
Glass Bottle
Catalog number 192130050
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
38.90
Save 5.25
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
38.90
Save 5.25
Each
Chemical Identifiers
CAS21679-31-2
IUPAC Namechromium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC15H21CrO6
InChI KeyJWORPXLMBPOPPU-LNTINUHCSA-K
SMILES[Cr+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
Appearance (Color)Purple
Elemental Analysis>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only