Hydroxylamine sulfate, 99%
Hydroxylamine sulfate, 99%
Hydroxylamine sulfate, 99%
Thermo Scientific Chemicals

Hydroxylamine sulfate, 99%

CAS: 10039-54-0 | H6N2O2·H2SO4 | 164.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 198530010
Price (USD)
119.65
Online exclusive
133.00
Save 13.35
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
119.65
Online exclusive
133.00
Save 13.35
Each
Chemical Identifiers
CAS10039-54-0
IUPAC Namebis(hydroxyazanium) sulfate
Molecular FormulaH8N2O6S
InChI KeyVGYYSIDKAKXZEE-UHFFFAOYSA-L
SMILES[NH3+]O.[NH3+]O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=98.5 %
Appearance (Color)White
Appearance (Form)Crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only