Xylenes, 96%, pure, mixed isomers with ethylbenzene
Xylenes, 96%, pure, mixed isomers with ethylbenzene
Xylenes, 96%, pure, mixed isomers with ethylbenzene
Xylenes, 96%, pure, mixed isomers with ethylbenzene
Thermo Scientific Chemicals

Xylenes, 96%, pure, mixed isomers with ethylbenzene

CAS: 1330-20-7 | C8H10
数量:
1 L
2.5 L
25 L
パッケージ:
すべて表示
Metal Drum
製品番号(カタログ番号) 200910250
価格(JPY)
-
見積もりを依頼する
数量:
25 L
パッケージ:
Metal Drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1330-20-7
IUPAC Name1,3-xylene
Molecular FormulaC8H10
InChI KeyIVSZLXZYQVIEFR-UHFFFAOYSA-N
SMILESCC1=CC(C)=CC=C1
さらに表示
Color scale=<20 APHA
Infrared spectrumConforms
GC>=95.0 % Xylene isomers + Ethylbenzene
Refractive index1.4940 to 1.5000 (20°C, 589 nm)
Appearance (Form)Clear liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only