Tolylene 2,4-diisocyanate, 75%, technical
Tolylene 2,4-diisocyanate, 75%, technical
Tolylene 2,4-diisocyanate, 75%, technical
Thermo Scientific Chemicals

Tolylene 2,4-diisocyanate, 75%, technical

CAS: 584-84-9 | C9H6N2O2 | 174.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 202500010
Price (USD)
130.65
Special offer
Online exclusive
Ends: 28-Dec-2026
153.00
Save 22.35
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
130.65
Special offer
Online exclusive
Ends: 28-Dec-2026
153.00
Save 22.35
Each
Chemical Identifiers
CAS584-84-9
IUPAC Name2,4-diisocyanato-1-methylbenzene
Molecular FormulaC9H6N2O2
InChI KeyDVKJHBMWWAPEIU-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1N=C=O)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.5660 to 1.5700 (20°C, 589 nm)
Appearance (Form)Liquid
NMR70 to 90 Tolylene 2,4-diisocyanate
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only