Sodium iodide, 99+%, pure, anhydrous
Sodium iodide, 99+%, pure, anhydrous
Sodium iodide, 99+%, pure, anhydrous
Thermo Scientific Chemicals

Sodium iodide, 99+%, pure, anhydrous

CAS: 7681-82-5 | INa | 149.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
2.5 kg
10 kg
Catalog number 203180100
Price (USD)
-
Quantity:
10 kg
Request bulk or custom format
Chemical Identifiers
CAS7681-82-5
IUPAC Namesodium iodide
Molecular FormulaINa
InChI KeyFVAUCKIRQBBSSJ-UHFFFAOYSA-M
SMILES[Na+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Loss on drying=<3 % (1 g, 105°C, 3 h)
Appearance (Form)Crystalline powder
Titration Argentometric>=99.0 % (on dried substance)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only