Pierce™ Protein A/G Plus Agarose, 2 mL - FAQs
Pierce™ Protein A/G Plus Agarose, 2 mL - FAQs
View additional product information for Pierce™ Protein A/G Plus Agarose - FAQs (20423, 20424)
1 product FAQs found
What does it mean that the crosslinked 6% beaded agarose is supplied as a 50% slurry for products Pierce Protein A/G Plus Agarose (Cat. No. 20423 and 20424) and Pierce Protein A/G Agarose (Cat. No. 20421 and 20422)?
This means that the solution consists of 50% settled resin and 50% liquid solution. For example, Cat. No. 20424 contains 20 ml of settled resin and 20 ml of storage buffer, making it a 40 ml, 50% slurry.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.