Florisil™, 100-200 mesh, for column chromatography
Florisil™, 100-200 mesh, for column chromatography
Florisil™, 100-200 mesh, for column chromatography
Thermo Scientific Chemicals

Florisil™, 100-200 mesh, for column chromatography

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
5 kg
Catalog number 205440050
Price (USD)
1,158.00
Each
Quantity:
5 kg
Request bulk or custom format
Price (USD)
1,158.00
Each
Chemical Identifiers
CAS1343-88-0
IUPAC Namesilicon(4+) magnesium(2+) trioxidandiide
Molecular FormulaMgO3Si
InChI KeyZADYMNAVLSWLEQ-UHFFFAOYSA-N
Molecular Weight (g/mol)100.39
View more
SpecificationsSpecification SheetSpecification Sheet
Particle size0.1 % (80 mesh)
Particle size46.0 % (140 mesh)
Particle size7.8 % (325 mesh)
Appearance (Form)Powder
Particle size7.5 % (100 mesh)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only