3,4-Diethoxybenzoic acid, 99%, Thermo Scientific™
3,4-Diethoxybenzoic acid, 99%, Thermo Scientific™
3,4-Diethoxybenzoic acid, 99%, Thermo Scientific™
Acros Organics

3,4-Diethoxybenzoic acid, 99%, Thermo Scientific™

 
Have Questions?
Catalog NumberQuantityPackaging
206170250
also known as 20617
25 gGlass bottle
Catalog number 206170250
also known as 20617
HSN 29189100
Price (INR)
15,284.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
15,284.00
Each
Chemical Identifiers
CAS5409-31-4
IUPAC Name3,4-diethoxybenzoic acid
Molecular FormulaC11H14O4
InChI KeyVVKCVAPLTRZJHH-UHFFFAOYSA-N
SMILESCCOC1=CC=C(C=C1OCC)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point167°C to 170°C
Appearance (Color)White to off-white
Infrared spectrumConforms
Titration with TBAH>=98.5 %
 
RUO – Research Use Only