Thermo Scientific Chemicals

Vanadium(V) oxide, 98+%, Thermo Scientific Chemicals

Catalog number: 207995000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Vanadium(V) oxide, 98+%, Thermo Scientific Chemicals

Catalog number: 207995000
500 g, Plastic bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
1314-62-1
IUPAC Name
[(dioxovanadio)oxy]dioxovanadium
Molecular Formula
O5V2
InChI Key
GNTDGMZSJNCJKK-UHFFFAOYSA-N
SMILES
O=[V](=O)O[V](=O)=O
Appearance (Form)
Powder
Sodium oxide (Na2O)
=<300 ppm
Iron (Fe)
=<1000 ppm
Chromium (Cr)
=<200 ppm
Assay
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text