Thermo Scientific Chemicals

Bismuth(III) oxide, 99.9%, extra pure, Thermo Scientific Chemicals

Catalog number: 208071000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Bismuth(III) oxide, 99.9%, extra pure, Thermo Scientific Chemicals

Catalog number: 208071000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 208071000
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
1304-76-3
IUPAC Name
dibismuth(3+) trioxidandiide
Molecular Formula
Bi2O3
InChI Key
TYIXMATWDRGMPF-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Bi+3].[Bi+3]
Appearance (Form)
Powder
Loss on drying
=<0.2 %
Bismuth (Bi)
89.5 to 89.9 %
Chloride (Cl)
=<10 ppm
Appearance (Color)
Yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text