L(+)-Isoleucine, 98.5-101.5%, USP, Multi-Compendial, GMP, J.T.Baker™
L(+)-Isoleucine, 98.5-101.5%, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

L(+)-Isoleucine, 98.5-101.5%, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, JP, BP and EP specifications.Leia mais
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoQuantity
2082-0712 kg
2082-05100 g
2082-061 kg
Número do catálogo 2082-07
Preço (BRL)
-
Requisite uma Cotação
Quantity:
12 kg
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
Especificações
CAS73-32-5
Chemical Name or MaterialL(+)-Isoleucine
ColorColorless
Melting Point279°C to 280°C
Molecular FormulaH3CCH2CH(CH3)CH(NH2)COOH
PackagingPolyethylene pail, screw top
Synonym(2S,3S)-(+)-Isoleucine, H-Ile-OH, L-Isoleucine
GradeMulti-Compendial/USP
Molecular Weight (g/mol)131.17
Percent Purity98.5 to 101.5%
Quantity12 kg
pH5.5 to 7
Unit SizeEach