Amberlite™ IR-120 Na ion-exchange resin
Amberlite™ IR-120 Na ion-exchange resin
Amberlite™ IR-120 Na ion-exchange resin
Thermo Scientific Chemicals

Amberlite™ IR-120 Na ion-exchange resin

CAS: 9002-23-7 | (C10H11)A(C8H9O3S)BC2H6
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 208412500
Price (USD)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS9002-23-7
SpecificationsSpecification SheetSpecification Sheet
Total volume capacity>=2 eq/l (Na+)
Harmonic mean size0.600 to 0.800 mm
Particle size=<2.0 % (< 0.300 mm)
Appearance (Color)Amber to brown
Moisture holding capacity45 to 50 % (Na+)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
RUO – Research Use Only