Specifications
CAS72-19-5
Chemischer Name oder MaterialL-Threonine
FarbeColorless
Schmelzpunkt255°C (Decomposes)
SummenformelC4H9NO3
VerpackungPoly bottle, tamper-evident cap
Synonym(2S,3R)-(-)-Threonine, (2S,3R)-2-Amino-3-hydroxybutyric acid, H-Thr-OH
GüteMulti-Kompendium/USP
Molekulargewicht (g/mol)119.12
Reinheit (%)98.5 to 101.5%
Menge1 kg
pH5 to 6.5
Unit SizeEach