5-Bromonicotinic acid, 98%
5-Bromonicotinic acid, 98%
5-Bromonicotinic acid, 98%
Thermo Scientific Chemicals

5-Bromonicotinic acid, 98%

CAS: 20826-04-4 | C6H4BrNO2 | 202.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 211390100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS20826-04-4
IUPAC Name5-bromopyridine-3-carboxylic acid
Molecular FormulaC6H4BrNO2
InChI KeyFQIUCPGDKPXSLL-UHFFFAOYSA-N
SMILESOC(=O)C1=CN=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration with TBAH>=97.5 %
Appearance (Color)White to yellow-grey to light brown
Melting point>=180°C
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only